4-Fluoro-2-methoxyphenol 97%
Supplier: Apollo Scientific
Warning
PC3817K-100G PC3817K-25G PC3817K-5G
APOSPC3817K-100GEA 272 EUR
APOSPC3817K-100G APOSPC3817K-25G APOSPC3817K-5G
4-Fluoro-2-methoxyphenol 97%
4-Fluoro-2-methoxyphenol
Formula: C₇H₇FO₂
MW: 142.13 g/mol
Boiling Pt: 195 °C
Density: 1.247 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00070797
CAS Number: 450-93-1