2-Fluoro-p-toluidine 99%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Fluoro-4-methylaniline, 4-Amino-3-fluorotoluene
PC3817KS-100G
PC3817KS-25G
PC3817KS-1G
APOSPC3817KS-100GEA
56.7
EUR
APOSPC3817KS-100G
APOSPC3817KS-25G
APOSPC3817KS-1G
2-Fluoro-p-toluidine 99%
2-Fluoro-p-toluidine
Formula:
C₇H₈FN MW: 125.15 g/mol Boiling Pt: 70…71 °C/7mmHg Melting Pt: 3 Density: 1.108 g/cm³ (25 °C) Flash Pt: 79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040975 CAS Number: 452-80-2 UN: 2810 ADR: 6.1 III |