3-Fluoro-o-toluidine 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-6-fluorotoluene, 3-Fluoro-2-methylaniline
PC3817L-100G PC3817L-25G PC3817L-250G PC3817L-500G
APOSPC3817L-100GEA 82.8 EUR
APOSPC3817L-100G APOSPC3817L-25G APOSPC3817L-250G APOSPC3817L-500G
3-Fluoro-o-toluidine 99%
3-Fluoro-o-toluidine
Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 205 °C
Melting Pt: 7 °C
Density: 1.099 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00007760
CAS Number: 443-86-7