3-Fluoro-p-toluidine 98%
Supplier: Apollo Scientific
Danger
PC3818-100G PC3818-25G
APOSPC3818-100GEA 78.3 EUR
APOSPC3818-100G APOSPC3818-25G
3-Fluoro-p-toluidine 98%
3-Fluoro-p-toluidine
Formula: C₇H₈FN
Boiling Pt: 93 °C/12mmHg
Melting Pt: 30…35 °C
Density: 1.093 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00007762
CAS Number: 452-77-7
EINECS: 207-212-4
UN: 2811
ADR: 6.1,III