4-Fluoro-m-toluidine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluoro-3-methylaniline, 5-Amino-2-fluorotoluene
PC3820-5G PC3820-100G PC3820-25G
APOSPC3820-5GEA 14.7 EUR
APOSPC3820-5G APOSPC3820-100G APOSPC3820-25G
4-Fluoro-m-toluidine 98%
4-Fluoro-m-toluidine
Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 85…86 °C/9mmHg
Melting Pt: 32…38 °C
Density: 1.12 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00025294
CAS Number: 452-69-7
EINECS: 207-207-7
UN: 2811
ADR: 6.1 III