5-Fluoro-o-toluidine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-4-fluorotoluene, 5-Fluoro-2-methylaniline
PC3822-100G PC3822-25G PC3822-500G
APOSPC3822-100GEA 27.7 EUR
APOSPC3822-100G APOSPC3822-25G APOSPC3822-500G
5-Fluoro-o-toluidine 98%
5-Fluoro-o-toluidine
Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 98…100 °C/15mmHg
Melting Pt: 37…40 °C
Density: 1.13
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007764
CAS Number: 367-29-3
EINECS: 206-689-6
UN: 2811
ADR: 6.1 III