3-Fluoro-p-toluic acid 97%
Supplier: Apollo Scientific
Warning
PC3823B-100G PC3823B-10G PC3823B-25G
APOSPC3823B-100GEA 55.1 EUR
APOSPC3823B-100G APOSPC3823B-10G APOSPC3823B-25G
3-Fluoro-p-toluic acid 97%
3-Fluoro-p-toluic acid
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00002490
CAS Number: 350-28-7
EINECS: 206-498-8