(±)-4-Fluoro-α-methylbenzylamine 96%
Supplier: Apollo Scientific
Danger
Synonyms: DL-4-Fluoro-α-methylbenzylamine, 1-(4-Fluorophenyl)ethylamine
PC3823CM-25G PC3823CM-50G PC3823CM-5G
APOSPC3823CM-25GEA 79.9 EUR
APOSPC3823CM-25G APOSPC3823CM-50G APOSPC3823CM-5G
(±)-4-Fluoro-α-methylbenzylamine 96%
(±)-4-Fluoro-α-methylbenzylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 76 °C/22mmHg
Density: 1.03 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00041323
CAS Number: 403-40-7
EINECS: 206-958-8
UN: 2735
ADR: 8 III