2-Fluoro-6-(4-methylphenoxy)benzonitrile 97%
Supplier: Apollo Scientific
Warning
PC3823R-50MG PC3823R-1G PC3823R-5G
APOSPC3823R-50MGEA 21.9 EUR
APOSPC3823R-50MG APOSPC3823R-1G APOSPC3823R-5G
2-Fluoro-6-(4-methylphenoxy)benzonitrile 97%
2-Fluoro-6-(4-methylphenoxy)benzonitrile
Formula: C₁₄H₁₀FNO
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00068203
CAS Number: 175204-08-7
UN: 3439
ADR: 6.1,III