2-Fluoro-4-methylpyridine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-4-picoline
PC3824-100G PC3824-25G
APOSPC3824-100GEA 48 EUR
APOSPC3824-100G APOSPC3824-25G
2-Fluoro-4-methylpyridine 97%
2-Fluoro-4-methylpyridine
Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 155…156 °C
Density: 1.078 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00041224
CAS Number: 461-87-0
UN: 1993
ADR: 3 III