2-Fluoro-6-methylpyridine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-6-picoline
PC3825-25G PC3825-5G PC3825-100G
APOSPC3825-25GEA 24.8 EUR
APOSPC3825-25G APOSPC3825-5G APOSPC3825-100G
2-Fluoro-6-methylpyridine 98%
2-Fluoro-6-methylpyridine
Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 140…141 °C
Density: 1.077 g/cm³ (25 °C)
Flash Pt: 66.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00041226
CAS Number: 407-22-7