5-(2-Fluorophenyl)-1H-tetrazole
Supplier: Apollo Scientific
Warning
Synonyms: 5-(2-Fluorophenyl)-1H-1,2,3,4-tetraazole
PC3845-1G PC3845-250MG
APOSPC3845-1GEA 29 EUR
APOSPC3845-1G APOSPC3845-250MG
5-(2-Fluorophenyl)-1H-tetrazole
5-(2-Fluorophenyl)-1H-tetrazole
Formula: C₇H₅FN₄
MW: 164.14 g/mol
Melting Pt: 157…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00798373
CAS Number: 50907-19-2