2-(2,3-Difluorophenyl)ethanol 98%
Supplier: Apollo Scientific
PC3857-1G PC3857-5G PC3857-10G
APOSPC3857-1GEA 66.7 EUR
APOSPC3857-1G APOSPC3857-5G APOSPC3857-10G
2-(2,3-Difluorophenyl)ethanol 98%
2-(2,3-Difluorophenyl)ethanol
Formula: C₈H₈F₂O
MW: 158.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862011
CAS Number: 126163-30-2