2-Fluoro-5-nitroaniline 98%
Supplier: Apollo Scientific
Warning
PC3860-100G PC3860-25G PC3860-5G
APOSPC3860-100GEA 55.1 EUR
APOSPC3860-100G APOSPC3860-25G APOSPC3860-5G
2-Fluoro-5-nitroaniline 98%
2-Fluoro-5-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 99…101 °C
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007652
CAS Number: 369-36-8
EINECS: 206-720-3
UN: 2811
ADR: 6.1,III