1-Fluoro-3-nitrobenzene 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Fluoronitrobenzene
PC3900-100G PC3900-25G
APOSPC3900-100GEA 0 EUR
APOSPC3900-100G APOSPC3900-25G
1-Fluoro-3-nitrobenzene 97%
1-Fluoro-3-nitrobenzene
Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 204…206 °C
Melting Pt: 2 °C
Density: 1.325 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00007196
CAS Number: 402-67-5
EINECS: 206-953-0
UN: 2810
ADR: 6.1 III