1-Fluoro-4-nitrobenzene 99%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Fluoronitrobenzene, 1-Fluoro-4-nitrobenzene
PC3910-250G PC3910-1KG
APOSPC3910-250GEA 43.5 EUR
APOSPC3910-250G APOSPC3910-1KG
1-Fluoro-4-nitrobenzene 99%
1-Fluoro-4-nitrobenzene
Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 205 °C
Melting Pt: 22…24 °C
Density: 1.33 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00007282
CAS Number: 350-46-9
EINECS: 206-502-8
UN: 2811
ADR: 6.1 III