5-Chloro-2,4-difluorobenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 5-Chloro-2,4-difluorobenzenesulphonylchloride
PC3921-5G PC3921-1G
APOSPC3921-5GEA 0 EUR
APOSPC3921-5G APOSPC3921-1G
5-Chloro-2,4-difluorobenzenesulphonyl chloride 97%
5-Chloro-2,4-difluorobenzenesulphonyl chloride
Formula: C₆H₂Cl₂F₂O₂S
MW: 247.05 g/mol
Melting Pt: 44…45 °C
Storage Temperature: Ambient
MDL Number: MFCD01940426
CAS Number: 13656-57-0
UN: 1759
ADR: 8 III