(Trifluoromethyl)trimethylsilane 0,5 M in THF 98%
Supplier: Apollo Scientific
Danger
Synonyms: trimethyl(trifluoromethyl)silane
PC3953-100ML PC3953-25ML PC3953-5ML
APOSPC3953-100MLEA 125 EUR
APOSPC3953-100ML APOSPC3953-25ML APOSPC3953-5ML
(Trifluoromethyl)trimethylsilane 0,5 M in THF 98%
(Trifluoromethyl)trimethylsilane
Formula: C₄H₉F₃Si
MW: 142.2 g/mol
Boiling Pt: 40 °C
Density: 0.895 g/cm³ (25 °C)
Flash Pt: –20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00145454
CAS Number: 81290-20-2
UN: 1993
ADR: 3 III