(R)-(+)-4-Fluoro-α-methylbenzylamine 97%
Supplier: Apollo Scientific
Danger
Synonyms: (R)-(+)-1-(4-Fluorophenyl)ethylamine, (R)-1-(4-Fluorophenyl)ethanamine
PC3962-1G PC3962-25G PC3962-5G
APOSPC3962-1GEA 24.8 EUR
APOSPC3962-1G APOSPC3962-25G APOSPC3962-5G
(R)-(+)-4-Fluoro-α-methylbenzylamine 97%
(R)-(+)-4-Fluoro-α-methylbenzylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 76 °C/22mmHg
Melting Pt: –30 °C
Density: 1.03 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03093090
CAS Number: 374898-01-8
UN: 2735
ADR: 8 II