2-Fluoro-4-nitrophenol 98%
Supplier: Apollo Scientific
Warning
PC3969-100G PC3969-25G PC3969-500G
APOSPC3969-100GEA 60.9 EUR
APOSPC3969-100G APOSPC3969-25G APOSPC3969-500G
2-Fluoro-4-nitrophenol 98%
2-Fluoro-4-nitrophenol
Formula: C₆H₄FNO₃
MW: 157.1 g/mol
Melting Pt: 120…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00051970
CAS Number: 403-19-0