1-(Difluoromethoxy)-3-nitrobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Difluoromethoxy)nitrobenzene
PC3971-100G PC3971-25G PC3971-5G
APOSPC3971-100GEA 376 EUR
APOSPC3971-100G APOSPC3971-25G APOSPC3971-5G
1-(Difluoromethoxy)-3-nitrobenzene 98%
1-(Difluoromethoxy)-3-nitrobenzene
Formula: C₇H₅F₂NO₃
MW: 189.12 g/mol
Boiling Pt: 240…242 °C
Density: 1.391 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD03407974
CAS Number: 22236-07-3