1-Chloro-2,4-bis(trifluoromethyl)benzene 97%
Supplier: Apollo Scientific
Warning
PC3987-100G PC3987-25G PC3987-5G
APOSPC3987-100GEA 319 EUR
APOSPC3987-100G APOSPC3987-25G APOSPC3987-5G
1-Chloro-2,4-bis(trifluoromethyl)benzene 97%
1-Chloro-2,4-bis(trifluoromethyl)benzene
Formula: C₈H₃ClF₆
Boiling Pt: 150…154 °C
Density: 1.52 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01631638
CAS Number: 327-76-4
UN: 1993
ADR: 3,III