trans-4-Fluoro-β-nitrostyrene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Fluoro-β-nitrostyrene
PC3988C-25G PC3988C-5G PC3988C-100G
APOSPC3988C-25GEA 114 EUR
APOSPC3988C-25G APOSPC3988C-5G APOSPC3988C-100G
trans-4-Fluoro-β-nitrostyrene 98%
trans-4-Fluoro-β-nitrostyrene
Formula: C₈H₆FNO₂
MW: 167.14 g/mol
Melting Pt: 99…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00024824
CAS Number: 706-08-1
EINECS: 211-891-2