4-(tert-Butoxy)-2,3,5,6-tetrafluorostyrene 97%
Supplier: Apollo Scientific
Warning
PC3989-5G PC3989-1G
APOSPC3989-5GEA 475 EUR
APOSPC3989-5G APOSPC3989-1G
4-(tert-Butoxy)-2,3,5,6-tetrafluorostyrene 97%
4-(tert-Butoxy)-2,3,5,6-tetrafluorostyrene
Formula: C₁₂H₁₂F₄O
Boiling Pt: 52 °C/ 0.04mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD04038273
CAS Number: 343305-41-9