4-Bromo-2-fluorobenzoic acid 98%
Supplier: Apollo Scientific
Warning
PC3994-100G PC3994-25G PC3994-5G
APOSPC3994-100GEA 69.6 EUR
APOSPC3994-100G APOSPC3994-25G APOSPC3994-5G
4-Bromo-2-fluorobenzoic acid 98%
4-Bromo-2-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 210…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00143263
CAS Number: 112704-79-7