3-(Trifluoromethoxy)benzenesulphonamide
Supplier: Apollo Scientific
Warning
PC4004-1G PC4004-25G PC4004-5G
APOSPC4004-1GEA 29 EUR
APOSPC4004-1G APOSPC4004-25G APOSPC4004-5G
3-(Trifluoromethoxy)benzenesulphonamide
3-(Trifluoromethoxy)benzenesulphonamide
Formula: C₇H₆F₃NO₃S
MW: 241.19 g/mol
Melting Pt: 100…101 °C
Storage Temperature: Ambient
MDL Number: MFCD03094233
CAS Number: 503179-70-2