2-(2-Aminopropan-2-yl)-N-(4-fluorobenzyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidine-4-carboxamide
Supplier: Apollo Scientific
PC400516-1G PC400516-5G
APOSPC400516-1GEA 41.6 EUR
APOSPC400516-1G APOSPC400516-5G
2-(2-Aminopropan-2-yl)-N-(4-fluorobenzyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidine-4-carboxamide
2-(2-Aminopropan-2-yl)-N-(4-fluorobenzyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidine-4-carboxamide
Formula: C₁₆H₁₉FN₄O₃
Storage Temperature: Ambient
MDL Number: MFCD10698741
CAS Number: 518048-03-8