1-Fluorocyclopropanecarboxylic acid 97%
Supplier: Apollo Scientific
Warning
PC400556-1G PC400556-250MG PC400556-5G
APOSPC400556-1GEA 109 EUR
APOSPC400556-1G APOSPC400556-250MG APOSPC400556-5G
1-Fluorocyclopropanecarboxylic acid 97%
1-Fluorocyclopropanecarboxylic acid
Formula: C₄H₅FO₂
MW: 104.08 g/mol
Melting Pt: 72…76 °C
Storage Temperature: Ambient
MDL Number: MFCD18205954
CAS Number: 137081-41-5