2-Chloro-6-methoxy-4-trifluoromethylpyridine
Supplier: Apollo Scientific
PC400636-1G PC400636-5G
APOSPC400636-1GEA 190 EUR
APOSPC400636-1G APOSPC400636-5G
2-Chloro-6-methoxy-4-trifluoromethylpyridine
2-Chloro-6-methoxy-4-trifluoromethylpyridine
Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845745
CAS Number: 1160994-99-9