4-Fluoro-3-nitroanisole
Supplier: Apollo Scientific
Warning
Synonyms: 1-Fluoro-4-methoxy-2-nitrobenzene
PC400727-1G PC400727-5G
APOSPC400727-1GEA 21.9 EUR
APOSPC400727-1G APOSPC400727-5G
4-Fluoro-3-nitroanisole
4-Fluoro-3-nitroanisole
Formula: C₇H₆FNO₃
MW: 171.13 g/mol
Melting Pt: 52…53 °C
Storage Temperature: Ambient
MDL Number: MFCD11848442
CAS Number: 61324-93-4