3-Fluoro-5-nitroaniline 95%
Supplier: Apollo Scientific
Warning
PC400728-1G PC400728-25G PC400728-5G
APOSPC400728-1GEA 134 EUR
APOSPC400728-1G APOSPC400728-25G APOSPC400728-5G
3-Fluoro-5-nitroaniline 95%
3-Fluoro-5-nitroaniline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 112…116 °C
Storage Temperature: Refrigerator
CAS Number: 2369-12-2