2,4-Difluoro-5-methylbenzonitrile 97%
Supplier: Apollo Scientific
PC400729-5G PC400729-1G PC400729-25G
APOSPC400729-5GEA 81.2 EUR
APOSPC400729-5G APOSPC400729-1G APOSPC400729-25G
2,4-Difluoro-5-methylbenzonitrile 97%
2,4-Difluoro-5-methylbenzonitrile
Formula: C₈H₅F₂N
MW: 153.13 g/mol
Melting Pt: 43…50 °C
Storage Temperature: Ambient
MDL Number: MFCD09835341
CAS Number: 329314-68-3