5-Bromo-3-chloro-2-fluoropyridine 98%
Supplier: Apollo Scientific
Warning
PC400730-25G
APOSPC400730-25GEA 85.7 EUR
APOSPC400730-25G
5-Bromo-3-chloro-2-fluoropyridine 98%
5-Bromo-3-chloro-2-fluoropyridine
Formula: C₅H₂BrClFN
MW: 210.43 g/mol
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD09834365
CAS Number: 38185-56-7