5'-Bromo-2'-fluoroacetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 1-(5-Bromo-2-fluorophenyl)ethanone
PC400732-100G PC400732-25G PC400732-5G
APOSPC400732-100GEA 259 EUR
APOSPC400732-100G APOSPC400732-25G APOSPC400732-5G
5'-Bromo-2'-fluoroacetophenone
5'-Bromo-2'-fluoroacetophenone
Formula: C₈H₆BrFO
MW: 217.04 g/mol
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD11226880
CAS Number: 198477-89-3