2-Fluoro-5-nitrotoluene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Fluoro-2-methyl-4-nitrobenzene
PC4010-100G PC4010-25G PC4010-500G
APOSPC4010-100GEA 24.8 EUR
APOSPC4010-100G APOSPC4010-25G APOSPC4010-500G
2-Fluoro-5-nitrotoluene 98%
2-Fluoro-5-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 98…101 °C/13mmHg
Melting Pt: 37…40 °C
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00007284
CAS Number: 455-88-9
EINECS: 207-251-7