2-Chloro-5-trifluoromethyl[1,3,4]thiadiazole 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-5-trifluoromethyl-[1,3,4]-thiadiazole
PC401017-5G
APOSPC401017-5GEA 434 EUR
APOSPC401017-5G
2-Chloro-5-trifluoromethyl[1,3,4]thiadiazole 95+%
2-Chloro-5-trifluoromethyl[1,3,4]thiadiazole
Formula: C₃ClF₃N₂S
MW: 188.56 g/mol
Boiling Pt: 64…66 °C/20mmHg
Storage Temperature: Ambient
MDL Number: MFCD04972766
CAS Number: 53645-98-0