2-Fluoro-6-nitrotoluene 98%
Supplier: Apollo Scientific
Warning
PC4011-100G PC4011-25G PC4011-500G
APOSPC4011-100GEA 46.4 EUR
APOSPC4011-100G APOSPC4011-25G APOSPC4011-500G
2-Fluoro-6-nitrotoluene 98%
2-Fluoro-6-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 208…210 °C
Melting Pt: 6.5…7 °C
Density: 1.27 g/cm³ (25 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00007197
CAS Number: 769-10-8
EINECS: 212-203-3