3-(Trifluoromethyl)styrene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(Trifluoromethyl)-3-vinylbenzene
PC4018-1G PC4018-25G PC4018-5G PC4018-10G
APOSPC4018-1GEA 30.6 EUR
APOSPC4018-1G APOSPC4018-25G APOSPC4018-5G APOSPC4018-10G
3-(Trifluoromethyl)styrene 97%
3-(Trifluoromethyl)styrene
Formula: C₉H₇F₃
MW: 172.15 g/mol
Boiling Pt: 64.5 °C/40mmHg
Density: 1.161 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075481
CAS Number: 402-24-4
UN: 1993
ADR: 3 III