3-Fluoro-4-nitrotoluene 99%
Supplier: Apollo Scientific
Warning
PC4020-100G PC4020-25G PC4020-5G
APOSPC4020-100GEA 60.9 EUR
APOSPC4020-100G APOSPC4020-25G APOSPC4020-5G
3-Fluoro-4-nitrotoluene 99%
3-Fluoro-4-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 97…98 °C/3mmHg
Melting Pt: 52…55 °C
Density: 1.438 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007053
CAS Number: 446-34-4
EINECS: 207-166-5