3-Amino-5-fluoro-6-methoxypyridine

Supplier: Apollo Scientific
Warning

Synonyms: 5-Fluoro-6-methoxypyridin-3-amine

PC402048-1G PC402048-100MG PC402048-250MG
APOSPC402048-1GEA 207 EUR
APOSPC402048-1G APOSPC402048-100MG APOSPC402048-250MG
3-Amino-5-fluoro-6-methoxypyridine
3-Amino-5-fluoro-6-methoxypyridine
Formula: C6H7FN2O
MW: 142.1300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07374960
CAS Number: 886372-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR