2-(Trifluoromethyl)benzoic acid hydrazide 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Trifluoromethyl)benzohydrazide, 2-(Trifluoromethyl)benzhydrazide
PC4021-1G PC4021-5G PC4021-10G
APOSPC4021-1GEA 27.7 EUR
APOSPC4021-1G APOSPC4021-5G APOSPC4021-10G
2-(Trifluoromethyl)benzoic acid hydrazide 95%
2-(Trifluoromethyl)benzoic acid hydrazide
Formula: C₈H₇F₃N₂O
MW: 204.150 g/mol
Melting Pt: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00221473
CAS Number: 344-95-6