3,5-Bis(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
Warning
PC4024-1G PC4024-10G PC4024-5G
APOSPC4024-1GEA 232 EUR
APOSPC4024-1G APOSPC4024-10G APOSPC4024-5G
3,5-Bis(trifluoromethyl)pyridine 97%
3,5-Bis(trifluoromethyl)pyridine
Formula: C₇H₃F₆N
MW: 215.1 g/mol
Boiling Pt: 117…118 °C
Melting Pt: 38…40 °C
Flash Pt: 44.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00128904
CAS Number: 20857-47-0
UN: 2926
ADR: 4.1+6.1,III