2,3-Bis(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Warning
PC4028-1G PC4028-5G
APOSPC4028-1GEA 108 EUR
APOSPC4028-1G APOSPC4028-5G
2,3-Bis(trifluoromethyl)pyridine
2,3-Bis(trifluoromethyl)pyridine
Formula: C₇H₃F₆N
MW: 215.1 g/mol
Boiling Pt: 85 °C/18mmHg
Melting Pt: –2 °C
Density: 1.5 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD03093763
CAS Number: 1644-68-4
UN: 2810
ADR: 6.1,III