N,N-Diethyl-2,2,2-trifluoroacetamide 99%
Supplier: Apollo Scientific
|
Warning
|
PC4048-25G
PC4048-5G
PC4048-100G
APOSPC4048-25GEA
73
EUR
APOSPC4048-25G
APOSPC4048-5G
APOSPC4048-100G
N,N-Diethyl-2,2,2-trifluoroacetamide 99%
N,N-Diethyl-2,2,2-trifluoroacetamide
Formula:
C₆H₁₀F₃NO Boiling Pt: 65…67 °C/24mmHg Density: 1.2 g/cm³ (25 °C) Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192034 CAS Number: 360-92-9 |