3-Fluoro-5-(pentafluorothio)benzyl bromide 98%
Supplier: Apollo Scientific
Danger
PC405659-1G PC405659-5G
APOSPC405659-1GEA 348 EUR
APOSPC405659-1G APOSPC405659-5G
3-Fluoro-5-(pentafluorothio)benzyl bromide 98%
3-Fluoro-5-(pentafluorothio)benzyl bromide
Formula: C₇H₅BrF₆S
MW: 315.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652459
CAS Number: 1240257-20-8
UN: 3265
ADR: 8,III