ω-Bromo-4-fluoroacetophenone 97%
Supplier: Apollo Scientific
Danger
PC4060-25G PC4060-5G PC4060-100G
APOSPC4060-25GEA 43.5 EUR
APOSPC4060-25G APOSPC4060-5G APOSPC4060-100G
ω-Bromo-4-fluoroacetophenone 97%
ω-Bromo-4-fluoroacetophenone
Intermediate for preparation of 1,2-diarylcyclopentenolcyclooxygenase inhibitors: J.Med.Chem.,37,3878,1994.
Formula: C₈H₆BrFO
Boiling Pt: 150 °C/12mmHg
Melting Pt: 45…49 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00040830
CAS Number: 403-29-2
EINECS: 206-955-1
UN: 3261
ADR: 8,II