1-Benzyl-3-bromo-5-fluoro-1H-1,2,4-triazole

Supplier: Apollo Scientific
Warning

PC4062-1G PC4062-5G
APOSPC4062-1GEA 132 EUR
APOSPC4062-1G APOSPC4062-5G
1-Benzyl-3-bromo-5-fluoro-1H-1,2,4-triazole
1-Benzyl-3-bromo-5-fluoro-1H-1,2,4-triazole
Formula: C₉H₇BrFN₃
Boiling Pt: 105 °C/0.3mmHg
Storage Temperature: Ambient
MDL Number: MFCD08435922
CAS Number: 214540-43-9
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR