2,2,2-Trichloro-1,1-bis(trifluoromethyl)ethanol 97%
Supplier: Apollo Scientific
PC4063-1G PC4063-5G
APOSPC4063-1GEA 107 EUR
APOSPC4063-1G APOSPC4063-5G
2,2,2-Trichloro-1,1-bis(trifluoromethyl)ethanol 97%
2,2,2-Trichloro-1,1-bis(trifluoromethyl)ethanol
Formula: C₄HCl₃F₆O
Boiling Pt: 139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236021
CAS Number: 7594-49-2