2-(4-Fluorophenyl)ethanol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluorophenethyl alcohol
PC4067-25G PC4067-5G
APOSPC4067-25GEA 71.2 EUR
APOSPC4067-25G APOSPC4067-5G
2-(4-Fluorophenyl)ethanol 97%
2-(4-Fluorophenyl)ethanol
Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 110 °C/20mmHg
Density: 1.121 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00002898
CAS Number: 7589-27-7
EINECS: 231-499-5