4-Fluorophenetole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluorophenyl ethyl ether, 1-Fluoro-4-ethoxybenzene, 1-Ethoxy-4-fluorobenzene
PC4068B-1G PC4068B-25G PC4068B-5G
APOSPC4068B-1GEA 21.9 EUR
APOSPC4068B-1G APOSPC4068B-25G APOSPC4068B-5G
4-Fluorophenetole 98%
4-Fluorophenetole
Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 91…93 °C/50mmHg
Melting Pt: –8.5 °C
Density: 1.0715 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00017953
CAS Number: 459-26-7
EINECS: 207-287-3